Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MafB Antibody (OTI1E9), Novus Biologicals™
SDP

Catalog No. NBP245718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245718 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245718 Supplier Novus Biologicals Supplier No. NBP245718
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry

MafB Monoclonal specifically detects MafB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MafB
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1E9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias Kreisler (mouse) maf-related leucine zipper homolog, Kreisler maf-related leucine zipper homolog, KRMLMAFB/Kreisler basic region/leucine zipper transcription factor, Maf-B, MGC43127, transcription factor MafB, V-maf musculoaponeurotic fibrosarcoma oncogene homolog B, v-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian)
Gene Symbols MAFB
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 32-320 of human MAFB (NP_005452) produced in E.coli.
Molecular Weight of Antigen 35.6 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9935
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.