Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MafG Antibody, Novus Biologicals™
SDP

Catalog No. NBP215019 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215019 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215019 Supplier Novus Biologicals Supplier No. NBP215019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MafG Polyclonal specifically detects MafG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MafG
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. O15525
Gene Alias basic leucine zipper transcription factor MafG, hMAF, MGC13090, MGC20149, transcription factor MafG, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein G, V-maf musculoaponeurotic fibrosarcoma oncogene homolog G, v-maf musculoaponeurotic fibrosarcoma oncogene homolog G (avian)
Gene Symbols MAFG
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MAFG. The exact sequence is proprietary.
Molecular Weight of Antigen 18 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4097
Test Specificity Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.