Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MafK Antibody (2F7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222570 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222570 Supplier Novus Biologicals Supplier No. NBP222570
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MafK Monoclonal specifically detects MafK in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MafK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:2000
Gene Accession No. NP_002351
Gene Alias FLJ32205, MGC71717, transcription factor MafK, ubiquitous (p18), v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian)
Gene Symbols MAFK
Host Species Mouse
Immunogen Recombinant human MAFK (1-156aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7975
Test Specificity Anti-human MAFK mAb, clone 2F7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MAFK protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.