Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MafK Antibody (2F7), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB310025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310025 Supplier Novus Biologicals Supplier No. NBP222570MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MafK Monoclonal antibody specifically detects MafK in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen MafK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F7
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias FLJ32205, MGC71717, transcription factor MafK, ubiquitous (p18), v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian)
Host Species Mouse
Immunogen Recombinant human MAFK (1-156aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7975
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.