Learn More
Description
Specifications
Specifications
| Antigen | MAGE 1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MA454 |
| Conjugate | Alexa Fluor 594 |
| Dilution | ELISA |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 |
| Gene Symbols | MAGEA1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
