Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169034
Description
MAGEA11 Polyclonal specifically detects MAGEA11 in Human samples. It is validated for Western Blot.Specifications
| MAGEA11 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MAGEA11 | |
| Synthetic peptides corresponding to MAGEA11 (melanoma antigen family A, 11) The peptide sequence was selected from the middle region of MAGEA11. Peptide sequence FSSTLNVGTLEELPAAESPSPPQSPQEESFSPTAMDAIFGSLSDEGSGSQ. | |
| Affinity purified | |
| RUO | |
| 4110 | |
| Human, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Cancer/testis antigen 1.11, CT1.11MAGE-11 antigen, MAGE11member 11, MAGEA-11, melanoma antigen family A, 11, melanoma-associated antigen 11, MGC10511 | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction