Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA3 Antibody (21B4), Allophycocyanin , Novus Biologicals™
SDP

Catalog No. NB233982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB233982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB233982 Supplier Novus Biologicals Supplier No. NBP320166APC
Add to Cart
Add to Cart

Human Monoclonal Antibody

MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen MAGEA3
Applications Flow Cytometry
Classification Monoclonal
Clone 21B4
Conjugate APC
Formulation PBS
Gene Alias Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613
Host Species Human
Immunogen The original antibody was isolated from a patient suffering from a tumor expressing a tumor specific antigen like MAGE-A3.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4102
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.