Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA3 Antibody (OTI1G9), PE, Novus Biologicals™
SDP

Catalog No. NB288016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288016 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288016 Supplier Novus Biologicals Supplier No. NBP272570PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen MAGEA3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1G9
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613
Host Species Mouse
Immunogen Full length human recombinant protein of human MAGEA3 (NP_005353) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4102
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.