Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA9 Antibody (OTI1D8), PE, Novus Biologicals™
SDP

Catalog No. NB288009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288009 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288009 Supplier Novus Biologicals Supplier No. NBP272572PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAGEA9 Monoclonal antibody specifically detects MAGEA9 in Human samples. It is validated for Western Blot

Specifications

Antigen MAGEA9
Applications Western Blot
Classification Monoclonal
Clone OTI1D8
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias Cancer/testis antigen 1.9, CT1.9MAGEA9B, MAGE-9 antigen, MAGE9cancer/testis antigen family 1, member 9, MAGEA9A, melanoma antigen family A, 9, melanoma-associated antigen 9, MGC8421
Host Species Mouse
Immunogen Full length human recombinant protein of human MAGEA9 (NP_005356) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4108
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.