Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEB1 Antibody (OTI4E12), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272573A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272573A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272573A405 Supplier Novus Biologicals Supplier No. NBP272573AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAGEB1 Monoclonal specifically detects MAGEB1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MAGEB1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4E12
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cancer/testis antigen 3.1, CT3.1cancer/testis antigen family 3, member 1, DAM10MAGE-B1 antigen, DSS/AHC critical interval MAGE superfamily 10, DSS-AHC critical interval MAGE superfamily 10, MAGEL1MAGE-XP antigen, MAGE-Xp, MAGEXP, melanoma antigen family B, 1, melanoma-associated antigen B1, MGC9322
Gene Symbols MAGEB1
Host Species Mouse
Immunogen Full length recombinant protein of human MAGEB1 (NP_796379) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4112.0
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.