Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGI3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP217210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217210 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217210 Supplier Novus Biologicals Supplier No. NBP217210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

MAGI3 Polyclonal specifically detects MAGI3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen MAGI3
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.94mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q5TCQ9
Gene Alias dJ730K3.2, MAGI-3, membrane associated guanylate kinase, WW and PDZ domain containing 3, membrane-associated guanylate kinase inverted 3, membrane-associated guanylate kinase-related 3, WW and PDZ domain-containing protein 3
Gene Symbols MAGI3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human MAGI3. The exact sequence is proprietary.
Molecular Weight of Antigen 166 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260425
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.