Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGI3 Sheep anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151719 250 μg
NB151720 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151719 Supplier Novus Biologicals Supplier No. NBP311883250UG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

MAGI3 Polyclonal antibody specifically detects MAGI3 in Human samples. It is validated for Western Blot

Specifications

Antigen MAGI3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 10 ug/ml
Formulation PBS
Gene Alias dJ730K3.2, MAGI-3, membrane associated guanylate kinase, WW and PDZ domain containing 3, membrane-associated guanylate kinase inverted 3, membrane-associated guanylate kinase-related 3, WW and PDZ domain-containing protein 3
Host Species Sheep
Immunogen Recombinant protein corresponding to the guanylate kinase-WW domain of MAGI3. (Uniprot: Q9H2V6)
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260425
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.