Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGI3 Sheep anti-Human, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody
$243.50 - $487.50
Specifications
Antigen | MAGI3 |
---|---|
Dilution | Western Blot 1 - 10 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MAGI3 Polyclonal antibody specifically detects MAGI3 in Human samples. It is validated for Western BlotSpecifications
MAGI3 | |
Western Blot | |
Unconjugated | |
Sheep | |
Human | |
dJ730K3.2, MAGI-3, membrane associated guanylate kinase, WW and PDZ domain containing 3, membrane-associated guanylate kinase inverted 3, membrane-associated guanylate kinase-related 3, WW and PDZ domain-containing protein 3 | |
Recombinant protein corresponding to the guanylate kinase-WW domain of MAGI3. (Uniprot: Q9H2V6) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1 - 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
260425 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title