Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagicMedia™ E. coli Expression Medium, Powder

Catalog No. p-7180343
Encompass_Preferred
Click to view available options
Quantity:
1 L
5 x 1 L

Includes

Contains a proprietary Component A (in foil pouch powder format), and Component B (in buffer liquid format).

MagicMedia E. coli Expression Medium is specifically designed to dramatically increase the yield of recombinant proteins in T7-regulated E. coli expression systems without monitoring culture OD (optical density) or adding inducer. The proprietary MagicMedia formulation enables E. coli growth to reach culture densities 3- to 10-fold higher than those achieved with traditional LB + IPTG methods, allowing you to:

• Achieve 3–10 times higher protein yields (Figure 1) with much less hands-on time
• Eliminate time-consuming OD monitoring steps—simply inoculate, grow the culture overnight, and harvest (Figure 2)
• Reduce culture volume to simplify handling and scale-up processes
• Ideal for simultaneous expression of multiple clones for high-throughput applications

MagicMedia E. coli Expression Medium, powder, consists of a proprietary Component A (in foil pouch powder format) and Component B (in buffer liquid format).

MagicMedia E. coli Expression Medium is also available in a ready-to-use, pre-sterilized liquid format (Cat. No. K6803).

Order Info

Store Component A (foil pouches) and Component B buffer (liquid) at Room Temperature (15 - 30°C).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Component A (foil pouches) and Component B (liquid) at room temperature (15–30°C).
Form Powder, Liquid
Format Kit
Product Type Microbiology Media
Product Line MagicMedia™
Quantity 1 L
Species Escherichia coli
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.