Magnesium iodide, White beads, 99.996% (Metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Manufacturer: Alfa Aesar 1428303

Quantity 1g

 View more versions of this product

Catalog No. AA1428303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10377-58-9
    Physical Form Beads
    Assay Percent Range 99.996% (Metals basis)
    Molecular Formula I2Mg
    Formula Weight 278.12
    MDL Number MFCD00049485
    Chemical Name or Material Magnesium iodide

    Specifications

    Physical Form Beads
    Formula Weight 278.12
    Quantity 1g
    Color White
    Density 4.43g/cm3 at 20°C
    Odor Odorless
    Sensitivity Light sensitive
    Assay Percent Range 99.996% (Metals basis)
    Molecular Formula I2Mg
    MDL Number MFCD00049485
    CAS 10377-58-9
    Chemical Name or Material Magnesium iodide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit