Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium titanium oxide, 99% (metals basis)

Catalog No. p-7048750
Click to view available options
Quantity:
250 g
1 kg

Catalog No. AA1139830

Supplier: Thermo Scientific Chemicals 01139830

Only null left
Add to Cart

CAS: 12032-30-3 | MgO3Ti | 120.169 g/mol

Magnesium titanium oxide is used in the primary chemical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-30-3
Molecular Formula MgO3Ti
Molecular Weight (g/mol) 120.169
MDL Number MFCD00016198
InChI Key XSETZKVZGUWPFM-UHFFFAOYSA-N
Synonym magnesium titanate, magnesium dititanate, karrooite, ticon 75, magnesium titanium trioxide, atd 16, magnesium titanate iv mgtio3, titanium magnesium oxide timgo3, magnesium dititanium pentoxide, magnesium titanate mgti2o5
PubChem CID 159407
IUPAC Name magnesium;oxygen(2-);titanium(4+)
SMILES [O-2].[O-2].[O-2].[Mg+2].[Ti+4]

Specifications

Melting Point 1610°C
Color White
Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 120.21
Percent Purity 99%
Odor Odorless
Density 3.36 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium titanium oxide
Show More Show Less
EINECSNumber 234-766-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.