Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium trifluoromethanesulfonate, 98%

Click to view available options
Quantity:
5g
25g
100g
60871-83-2
C2F6MgO6S2
322.431
MFCD00042601
BZQRBEVTLZHKEA-UHFFFAOYSA-L
magnesium trifluoromethanesulfonate,magnesium triflate,magnesium 2+ ditriflate,trifluoromethanesulfonic acid magnesium salt,magnesium trifluoromethanesulphonate,methanesulfonic acid, trifluoro-, magnesium salt,magnesiumtrifluoromethanesulfonate,magnesium trifiate,acmc-209mmv,ksc352a4p
2734125
magnesium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]
60871-83-2
C2F6MgO6S2
322.431
MFCD00042601
BZQRBEVTLZHKEA-UHFFFAOYSA-L
magnesium trifluoromethanesulfonate,magnesium triflate,magnesium 2+ ditriflate,trifluoromethanesulfonic acid magnesium salt,magnesium trifluoromethanesulphonate,methanesulfonic acid, trifluoro-, magnesium salt,magnesiumtrifluoromethanesulfonate,magnesium trifiate,acmc-209mmv,ksc352a4p
2734125
magnesium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]
Applications
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Sensitivity Hygroscopic
Melting Point >310°C
Quantity 100g
Linear Formula Mg(CF3SO3)2
UN Number UN1759
Beilstein 705578
Solubility Information Soluble in water.
Formula Weight 322.43
Percent Purity 98%
Chemical Name or Material Magnesium trifluoromethanesulfonate

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures