Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Magnetic Antibody Labeling Kit
SDP

Catalog No. p-7112401 Shop All R&D Systems Products
Click to view available options
Quantity:
200 μg
60 μg

Catalog No. 13-000-005

Supplier: Novus Biologicals™ 13000005

Allows one-step conjugation of antibodies, proteins and peptides.

  • Conjugates to specially treated magnetic particles
  • No extensive optimization of the conjugation reaction
  • Covalent attachment of antibodies to magnetic particles
  • Bead size: 0.5μm
  • Minimizes risk of unspecific binding

Applications: Immunoprecipitation experiments
Storage: Store at 4°C

Note: Kits are guaranteed for 6 months from date of receipt

Specifications

Quantity 60 μg
Product Type Magnetic antibody labeling kit

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.