Learn More
Description
Specifications
Specifications
| Antigen | MAGT1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | bA217H1.1, DKFZp564K142, FLJ14726, IAG2, IAPPRO0756, Implantation-associated protein, magnesium transporter 1, magnesium transporter protein 1, MagT1, MGC64926, MRX95, oligosaccharyltransferase 3 homolog B, OST3B |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 60-220 of human MAGT1 (NP_115497.4). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
