Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Major allergen I polypeptide chain 2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223611 100 μg
NBP22361150 50 μg
NB019355 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP223611 Supplier Novus Biologicals Supplier No. NBP223611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Major allergen I polypeptide chain 2 Polyclonal antibody specifically detects Major allergen I polypeptide chain 2 in Human, Mouse, Rat, Feline, Primate samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Major allergen I polypeptide chain 2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Accession No. P30440
Gene Alias AG4, allergen Cat-1, allergen FdI, allergen Fel d I-B, CH2, CH2 major allergen I, polypeptide chain 2, CH2 major allergen I, polypeptide chain 2 [ Felis catus (domestic cat) ], Fel d I, major allergen I, polypeptide chain 2
Gene Symbols CH2
Host Species Rabbit
Immunogen Synthetic peptides unique to Fel D1 protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 677879
Target Species Human, Mouse, Rat, Feline, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.