Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAK10 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB356724 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356724 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356724 Supplier Novus Biologicals Supplier No. NBP177056MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAK10 Polyclonal antibody specifically detects MAK10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MAK10
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias bA379P1.1, corneal wound healing-related protein, EGAP, embryonic growth-associated protein homolog, FLJ21613, FLJ22643, MAK10, MAK10 homolog, amino-acid N-acetyltransferase subunit, N(alpha)-acetyltransferase 35, NatC auxiliary subunit, N-alpha-acetyltransferase 35, NatC auxiliary subunit, protein MAK10 homolog
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human MAK10. The immunogen is located within amino acids 190 - 240 of MAK10. Amino Acid Squence: KDVEDDMQRRVKSTRSRQGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60560
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.