Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malondialdehyde Antibody (11E3), FITC, Novus Biologicals™
SDP

Catalog No. NBP259367F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259367F 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259367F Supplier Novus Biologicals Supplier No. NBP259367F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Malondialdehyde Monoclonal specifically detects Malondialdehyde in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Malondialdehyde
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 11000
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.