Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MALRD1 Antibody, Novus Biologicals™
SDP

Catalog No. NB437809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB437809 25ul
NBP214578 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB437809 Supplier Novus Biologicals Supplier No. NBP21457825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MALRD1 Polyclonal specifically detects MALRD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen C10orf112
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias bA265G8.2, C10orf112 chromosome 10 open reading frame 112, chromosome 10 open reading frame 112
Gene Symbols C10orf112
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: DSTVIWRVLYNQGKQWLEATIQLGRLSQPFHLSLDKVSLGIYDGVSAIDDIRFENCTLPLPAESCEGLDHFWCRHTRACIEKLRLCD
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340895
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.