Learn More
Description
Specifications
Specifications
| Antigen | MALT1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MT1/410 |
| Conjugate | Alexa Fluor 647 |
| Dilution | ELISA |
| Gene Alias | caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase |
| Gene Symbols | MALT1 |
| Host Species | Mouse |
| Immunogen | Human MALT1 recombinant fragment (aa701-808) (Uniprot: Q9UDY8) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
