Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MALT1 Antibody (rMT1/410), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB384237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384237 Supplier Novus Biologicals Supplier No. NBP308824AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MALT1 Monoclonal antibody specifically detects MALT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MALT1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMT1/410
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase
Host Species Mouse
Immunogen Recombinant fragment (around aa701-808) of human MALT1 protein (exact sequence is proprietary) (Uniprot: Q9UDY8)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10892
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.