Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MALT1 Antibody (SPM578), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB006873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB006873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB006873 Supplier Novus Biologicals Supplier No. NBP234811AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MALT1 Monoclonal specifically detects MALT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MALT1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM578
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase
Gene Symbols MALT1
Host Species Mouse
Immunogen Human MALT1 recombinant fragment (aa701-808) (Uniprot: Q9UDY8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Test Specificity Mucosa associated lymphoid tissue lymphoma translocation gene 1 (MALT1) is found in extranodal low-grade B cell lymphomas. MALT1 encodes two Ig-like C2-type domains and fuses with an API2 gene, which is highly expressed in adult lymphoid tissue. The translocation of this MALT1 gene, which maps to human chromosome 18q21, and the apoptosis-inhibiting API2 gene results in an increased development of MALT lymphomas and apoptosis inhibition. Sites at which this API2-MALT1 (11;18)(q21;q21) translocation commonly occurs are within human lung and kidney tissue. MALT lymphoma expresses nuclear Bcl10, which mediates the oligomerization and activation of a MALT1 caspase-like domain. MALT1 mRNA is found in pre-B cells, mature B cells and plasma cells.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.