Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MALT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102587 Supplier Invitrogen™ Supplier No. PA5102587
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MALT1.

MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MALT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MALT1
Gene Accession No. Q2TBA3, Q9UDY8
Gene Alias A630046N12; caspase-like protein; D430033E09Rik; DKFZp434L132; IMD12; MALT lymphoma-associated translocation; Malt1; malt-1; MALT1 paracaspase; MALT1 protease; MLT; MLT1; mucosa associated lymphoid tissue lymphoma translocation gene 1; mucosa-associated lymphoid tissue lymphoma translocation protein 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1 homolog; paracaspase; paracaspase 1; paracaspase-1; PCASP1
Gene Symbols MALT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MALT1(Accession Q9UDY8), corresponding to amino acid residues C317-Y367.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10892, 240354, 307366
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.