Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Maltitol, 95%

Catalog No. AC295800010
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle

Catalog No. AC295800010

Supplier: Thermo Scientific Chemicals 295800010

Only null left
Add to Cart

CAS: 585-88-6 | C12H24O11 | 344.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-88-6
Molecular Formula C12H24O11
Molecular Weight (g/mol) 344.32
MDL Number MFCD00006600
InChI Key VQHSOMBJVWLPSR-WUJBLJFYSA-N
Synonym maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk
PubChem CID 493591
ChEBI CHEBI:68428
IUPAC Name (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O

Specifications

Melting Point 147°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 kg
Beilstein 17, V,7, 145
Specific Rotation Condition + 107.00 (20.00°C c=5,water)
Specific Rotation + 107
Solubility Information (10% in water) Clear colorless
Formula Weight 344.32
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Maltitol, 95%
Show More Show Less
EINECSNumber 209-567-0
RTECSNumber LZ4394000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.