Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Antibody (10F2), Novus Biologicals™
SDP

Catalog No. NB298664 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298664 50 μL
NB298665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298664 Supplier Novus Biologicals Supplier No. NBP32615650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in Human, E. coli samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Maltose Binding Protein
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 10F2
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Host Species Rabbit
Immunogen A synthesized peptide derived from Escherichia coli Maltose Binding Protein [UniProt P0AEX9]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Epitope Tags, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Target Species Human, E. coli
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.