Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Antibody (10F2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Maltose Binding Protein |
---|---|
Clone | 10F2 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in Human, E. coli samples. It is validated for Flow Cytometry, ELISASpecifications
Maltose Binding Protein | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, E. coli | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
IgG | |
Affinity purified |
10F2 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Epitope Tags, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
A synthesized peptide derived from Escherichia coli Maltose Binding Protein [UniProt P0AEX9] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title