Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Antibody (R29.6), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB386933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386933 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386933 Supplier Novus Biologicals Supplier No. NBP308444AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in E. coli samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Maltose Binding Protein
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone R29.6
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Host Species Mouse
Immunogen MOS maltose binding protein fusion protein.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epitope Tags, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.