Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Antibody (R29.6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NP27661JF49 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27661JF49 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27661JF49 Supplier Novus Biologicals Supplier No. NBP276651JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Maltose Binding Protein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone R29.6
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Host Species Mouse
Immunogen MOS maltose binding protein fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This antibody is useful for detection and isolation of recombinant MBP fusion proteins.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.