Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Antibody (R29), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB166609AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166609AI 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB166609AI Supplier Novus Biologicals Supplier No. NB10066609AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Bacteria samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Maltose Binding Protein
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone R29
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Gene Symbols malE
Host Species Mouse
Immunogen Recombinant maltose binding protein
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 948538
Test Specificity Recognizes maltose binding protein (MBP). The is useful in recognizing proteins expressed with an MBP tag.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.