Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Antibody (TVG MBP 3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276653
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| Maltose Binding Protein | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot : 1:100 - 1:2000, ELISA, Immunoprecipitation : 1:10 - 1:500 | |
| Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
| A single intravenous injection of the maltose binding protein MBP-E2. | |
| 0.1 mL | |
| This antibody is useful for detection and isolation of recombinant MBP fusion proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, ELISA, Immunoprecipitation | |
| TVG MBP 3 | |
| Unconjugated | |
| PBS with 0.02% sodium azide | |
| Mouse | |
| Thiophilic Chromatography | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction