Learn More
Invitrogen™ Maltose Binding Protein Polyclonal Antibody

Description
PA1-989 detects recombinant maltose binding protein. PA1-989 has been used successfully in Western blotting to detect the presence of maltose binding protein tagged proteins. The PA1-989 immunogen is recombinant maltose binding protein.
Specifications
Specifications
| Antigen | Maltose Binding Protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene Alias | MBP; MBP Tag |
| Host Species | Rabbit |
| Immunogen | Recombinant maltose binding protein. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.