Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML2 Antibody (MAML2/1302), Novus Biologicals™
SDP

Catalog No. NB253228100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253228100 100 μg
NB25322820 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB253228100 Supplier Novus Biologicals Supplier No. NBP253228100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAML2 Monoclonal specifically detects MAML2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MAML2
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAML2/1302
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias DKFZp686N0150, KIAA1819MGC176701, mam-2, MAM2, MAM-3, MAM3MLL-MAML2, mastermind (Drosophila)-like 2, mastermind-like 2 (Drosophila), mastermind-like protein 2
Gene Symbols MAML2
Host Species Mouse
Immunogen Recombinant human full-length MAML2
Molecular Weight of Antigen 125 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84441
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.