Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML3 Antibody (MAML3/1303), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$203.51 - $480.74

Specifications

Antigen MAML3
Clone MAML3/1303
Dilution Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, SDS-Page, Immunofluorescence 0.5-1ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP247646F
SDP
View Documents
Novus Biologicals
NBP2476460.02MG
0.02 mg
Each for $203.51
Only null left
Add to Cart
 
NBP247646A
SDP
View Documents
Novus Biologicals
NBP2476460.1MG
0.1 mg
Each for $480.74
Only null left
Add to Cart
 
NBP247646B
SDP
View Documents
Novus Biologicals
NBP2476460.2MG
0.2 mg N/A N/A N/A
Description

Description

MAML3 Monoclonal specifically detects MAML3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MAML3
Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, SDS-Page, Immunofluorescence 0.5-1ug/ml
Monoclonal
Purified
RUO
Human
CAG/CTG 3, ERDA3, MAM-2, mam-3, mastermind-like 3 (Drosophila), mastermind-like protein 3, polyglutamine rich, trinucleotide repeat containing 3
MAML3
IgG1
Protein A or G purified
MAML3 (mastermind-like protein 3) is a nuclear speckle protein that acts as a transcriptional coactivator for Notch receptors. The Notch signaling pathway influences cell fate by regulating the ability of precursor cells to properly respond to developmental signals. MAML3 is a member of the mastermind-like family of proteins that are human homologs of the Drosophila melanogaster mastermind protein. Through its N-terminal region, MAML3 interacts with the ankyrin repeats of the Notch proteins Notch 1, Notch 2, Notch 3 and Notch 4. This interaction leads to formation of a DNA-binding complex with the Notch proteins and RBP-Jk; a complex that can then induce HES1 gene expression. While the N-terminal domain of MAML3 is essential for proper Notch binding, the C-terminal domain of MAML3 is essential for transcriptional activation. Due to its involvement in cell signaling and transcriptional activation, upregulation of MAML3 is thought to be involved in oncogenesis.
MAML3/1303
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Unconjugated
Mouse
Neuroscience
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
55534
Recombinant human MAML3 protein fragment
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.