Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mammaglobin A Antibody (MGB/4058R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB391898 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391898 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391898 Supplier Novus Biologicals Supplier No. NBP308586JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Mammaglobin A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MGB/4058R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1
Host Species Rabbit
Immunogen Recombinant full-length human Mammaglobin A protein (Uniprot: Q13296)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4250
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.