Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mammaglobin A Mouse anti-Human, Clone: MGB/2704, Novus Biologicals™
SDP

Catalog No. NB122056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122056 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122056 Supplier Novus Biologicals Supplier No. NBP30729120UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen Mammaglobin A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MGB/2704
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1
Host Species Mouse
Immunogen Recombinant full-length human Mammaglobin A protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4250
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.