Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAMSTR Antibody, Novus Biologicals™
SDP

Catalog No. NBP16920420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16920420 20 μL
NBP169204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16920420 Supplier Novus Biologicals Supplier No. NBP16920420UL

Rabbit Polyclonal Antibody

MAMSTR Polyclonal specifically detects MAMSTR in Human samples. It is validated for Western Blot.

Specifications

Antigen MAMSTR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Alias FLJ36070, MASTRlikely ortholog of MEF2-activating SAP transcriptional regulator, MEF2 activating motif and SAP domain containing transcriptional regulator, MEF2-activating motif and SAP domain-containing transcriptional regulator, MEF2-activating SAP transcriptional regulator, MEF2-activating SAP transcriptional regulatory protein, MGC117259
Gene Symbols MAMSTR
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FLJ36070 (MEF2 activating motif and SAP domain containing transcriptional regulator) The peptide sequence was selected from the middle region of FLJ36070. Peptide sequence PRAADTPGTAPAPTPTPAPAAAPALTPSSGP
Molecular Weight of Antigen 33 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284358
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.