Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAMSTR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | MAMSTR |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP169204
![]() |
Novus Biologicals
NBP169204 |
100 μL |
Each for $487.50
|
|
|||||
NBP16920420
![]() |
Novus Biologicals
NBP16920420UL |
20 μL | N/A | N/A | N/A | ||||
Description
MAMSTR Polyclonal specifically detects MAMSTR in Human samples. It is validated for Western Blot.Specifications
MAMSTR | |
Polyclonal | |
Rabbit | |
FLJ36070, MASTRlikely ortholog of MEF2-activating SAP transcriptional regulator, MEF2 activating motif and SAP domain containing transcriptional regulator, MEF2-activating motif and SAP domain-containing transcriptional regulator, MEF2-activating SAP transcriptional regulator, MEF2-activating SAP transcriptional regulatory protein, MGC117259 | |
MAMSTR | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
284358 | |
IgG | |
33 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title