Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) iodide, anhydrous, 98%

Catalog No. p-7049994
Click to view available options
Quantity:
10 g
50 g

CAS: 7790-33-2

Manganese(II) iodide can be used as a pink pigment or as a source of the manganese ion or iodide ion. It is often used in the lighting industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-33-2
MDL Number MFCD00016225

Specifications

Melting Point 638°C
Color Red to White
Physical Form Powder
Quantity 10 g
Linear Formula MnI2
Merck Index 14,5732
Solubility Information Soluble in water with gradual decomposition; Soluble in alcohol
Formula Weight 308.75
Percent Purity 98%
Odor Odorless
Sensitivity Air, light, and moisture sensitive
Density 5.0 g/mL
Chemical Name or Material Manganese(II) iodide, Anhydrous
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-201-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.