Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(III) acetate hydrate, 96%

Catalog No. p-7049871
Click to view available options
Quantity:
10 g
50 g

CAS: 19513-05-4 | C6H9MnO6 | 232.07 g/mol

It acts as a mild and selective oxidizing agent in organic synthesis and materials science. Manganese(III) Acetate Dihydrate is also a reagent used for radical cyclizations and α-keto-acetoxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19513-05-4
Molecular Formula C6H9MnO6
Molecular Weight (g/mol) 232.07
MDL Number MFCD00150022
InChI Key AHSBSUVHXDIAEY-UHFFFAOYSA-K
IUPAC Name manganese(3+) triacetate
SMILES [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color Pink
Physical Form Crystalline
Quantity 10 g
Linear Formula Mn(OOCCH3)3·xH2O
Solubility Information It is soluble in water.
Formula Weight 232.07 (Anhydrous)
Percent Purity 96%
Sensitivity Hygroscopic
Density 1.59 g/mL
Chemical Name or Material Manganese(III) acetate hydrate
Hazard Category H315-H319-H335-H336-H373
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
EINECSNumber 213-602-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.