Learn More
Manganese(III) fluoride, 98% (metals basis)
Description
It is used as a fluorinating agent, useful for conversion of hydrocarbons into fluorocarbons. It is used in organic synthesis as Lewis acid to prepare a number of organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Physical Form | Crystalline |
| Quantity | 10 g |
| UN Number | UN3288 |
| Merck Index | 14,5741 |
| Solubility Information | Insoluble in organic solvents. Hydrolyzed by water. |
| Formula Weight | 111.93 |
| Percent Purity | 98% |
| Sensitivity | Hygroscopic |
| Density | 3.54 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H272-H302+H332-H315-H318-H335 |
| Hazard Statement | GHS H Statement H331-H302-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: II |
| EINECSNumber | 232-006-6 |
| RTECSNumber | OP0882600 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.