Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mannitol, BiotechGrade, 96-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604222 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-222 2.5 kg Poly Bottle
18-604-221 500 g Poly Bottle
18-604-223 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-222 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M11282.5KG
Add to Cart
Add to Cart

M1128, 69-65-8, C6H14O6

Spectrum™ Chemical Mannitol, BiotechGrade

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Melting Range 164-169°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Rotation +137° to +145°
Percent Purity 96 to 101.5%
Grade Biotech
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Mannitol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.