Learn More
Eisco™ Manual Spirometry Lung Volume Kit and Replacement Parts
SCI_ED
Catalog No. p-7230412 Shop All Eisco Products
Change view
Click to view available options
Product Type:
Manual Spirometry Set
Replacement Mouthpieces
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S15103 Manual Spirometry Set
S15104 Replacement Mouthpieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15103 Supplier Eisco™ Supplier No. LNGKIT
Only null left
Add to Cart
Add to Cart
Manual Spirometry Lung Volume Set includes fourteen pieces that can be used to study the volume of gas in the lungs during the respiratory cycle.
  • Useful for studying the various factors that can change a person's lung capacity such as old age, diaphragm weakness, and loss of elasticity
  • Average lung capacity is approximately 6 liters
  • Volumes will vary depending on the depth of respiration, ethnicity, gender, age, body composition, and in the presence of certain respiratory diseases
  • Kit includes 4 mouthpieces, 4 plastic bags calibrated to 6 L, and 6 elastic mouthpiece holders
  • Mouthpieces can be washed and disinfected
  • For educational or demonstration use only
  • Do not use to diagnose any medical conditions
Specifications
Product Type Manual Spirometry Set
For Use With (Application) Studying Lung Capacity
Includes 4 Mouthpieces, 4 Plastic Bags Calibrated to 6 L, and 6 Elastic Mouthpiece Holders