Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1B Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10068256R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068256R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068256R Supplier Novus Biologicals Supplier No. NB10068256R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP1B Polyclonal specifically detects MAP1B in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen MAP1B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP-1B, MAP5, microtubule-associated protein 1B
Gene Symbols MAP1B
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 900 and 950 of human microtubule-associated protein 1B using the numbering given in entry NP_005900.2 (GeneID 4131).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4131
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.