Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1B, p Thr1265 Antibody, HRP - SuperBUGS, Novus Biologicals™
SDP

Catalog No. NBP142827H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142827H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142827H Supplier Novus Biologicals Supplier No. NBP142827H

Rabbit Polyclonal Antibody

MAP1B Polyclonal specifically detects MAP1B in Human, Mouse, Rat, Chicken, Fish, Monkey, Reptile, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen MAP1B (p Thr1265)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP-1B, MAP5, microtubule-associated protein 1B
Gene Symbols MAP1B
Host Species Rabbit
Immunogen Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot# P15205]
Molecular Weight of Antigen 320 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4131
Test Specificity Phosphorylated Thr1265 on MAP1B (Thr1261 in rat).
Target Species Human, Mouse, Rat, Chicken, Fish, Monkey, Reptile, Xenopus species
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.