Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1S Antibody, Novus Biologicals™
SDP

Catalog No. NBP147281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147281 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147281 Supplier Novus Biologicals Supplier No. NBP147281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP1S Polyclonal specifically detects MAP1S in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MAP1S
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias BPY2 interacting protein 1, BPY2IP1VCY2IP-1, C19orf5, chromosome 19 open reading frame 5, FLJ10669, MAP8MAP-1S, MGC133087, microtubule-associated protein 1S, Microtubule-associated protein 8, Variable charge Y chromosome 2-interacting protein 1, VCY2 interacting protein 1, VCY2-interacting protein 1, VCY2IP1BPY2-interacting protein 1
Gene Symbols MAP1S
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 475 and 525 of human MAP1S using the numbering given in entry NP_060644.4 (GeneID 55201).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55201
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.