Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Antibody (2C4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB362579 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362579 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362579 Supplier Novus Biologicals Supplier No. NBP250032IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MAP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2C4
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Host Species Mouse
Immunogen MAP2 Antibody (2C4) was developed against full length recombinant human MAP2D. [UniProt# P11137]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4133
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.